Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEPTOR/DEPDC6 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 13 publications
$206.50 - $499.50
Specifications
| Antigen | DEPTOR/DEPDC6 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP149674SS
![]() |
Novus Biologicals
NBP149674SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP149674
![]() |
Novus Biologicals
NBP149674 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
DEPTOR/DEPDC6 Polyclonal specifically detects DEPTOR/DEPDC6 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DEPTOR/DEPDC6 | |
| Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| DEP domain containing 6, DEP domain containing mTOR interacting protein, DEP domain containing MTOR-interacting protein, DEP domain-containing mTOR-interacting protein, DEP domain-containing protein 6, DEP.6, DEPDC6FLJ12341, DKFZp564B1778, FLJ12428, FLJ13854 | |
| DEPTOR | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Predicted to react with bovine based on 100% sequence homology. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Bovine | |
| Q8TB45, Q570Y9 | |
| 64798 | |
| A genomic peptide made to an internal region of the human DEPTOR protein (within residues 10-200). [Swiss-Prot: Q8TB45] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title