Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DERA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565986 Supplier Invitrogen™ Supplier No. PA565986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISKIQVNHPAV LRRAEQIQAR RTVKKEWQAA WLLKAVTFID LTTLSGDDTS SNIQRLCYKA KYPIREDLLK ALNMHDKGIT TAAVCVYPAR VCDAVKALKA AGCNIPVAS Highest antigen sequence identity to the following orthologs - mouse 92%, rat 91%.

Catalyzes a reversible aldol reaction between acetaldehyde and D-glyceraldehyde 3-phosphate to generate 2-deoxy-D-ribose 5-phosphate. Participates in stress granule (SG) assembly. May allow ATP production from extracellular deoxyinosine in conditions of energy deprivation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DERA
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DERA
Gene Accession No. Q9Y315
Gene Alias 2-deoxy-D-ribose 5-phosphate aldolase; 2-deoxyribose-5-phosphate aldolase homolog; CGI-26; DEOC; deoxyriboaldolase; Deoxyribose-phosphate aldolase; deoxyribose-phosphate aldolase (putative); DERA; Phosphodeoxyriboaldolase; putative deoxyribose-phosphate aldolase
Gene Symbols DERA
Host Species Rabbit
Immunogen Recombinant Human DERA. Recombinant protein control fragment (Product #RP-105030).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51071
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.