Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DERF2 Antibody (73) [CoraFluor™ 1], Novus Biologicals™
SDP

Catalog No. NB777380 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB777380 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB777380 Supplier Novus Biologicals Supplier No. NBP348429CL1
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DERF2 Monoclonal antibody specifically detects DERF2 in Invertebrate samples. It is validated for ELISA

Specifications

Antigen DERF2
Applications ELISA
Classification Monoclonal
Clone 73
Conjugate CoraFluor 1
Formulation PBS
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dermatophagoides farinae DERF2.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Invertebrate
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.