Learn More
Invitrogen™ DERL2 Polyclonal Antibody

Description
DERL2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | DERL2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | DERL2 |
| Gene Accession No. | Q8BNI4 |
| Gene Alias | carcinoma related; CGI-101; Degradation in endoplasmic reticulum protein 2; Der1-like domain family, member 2; der1-like protein 2; Der2; Derl2; derlin 2; derlin-2; derlin-2-like; DERtrin-2; F-LAN-1; Flana; F-LANa; LOC100910823; SBBI53 |
| Gene Symbols | DERL2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.