Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DERL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542574 Supplier Invitrogen™ Supplier No. PA542574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FFFNMLFVFR YCRMLEEGSF RGRTADFVFM FLFGGVLMTL LGLLGSLFFL Sequence homology: Cow: 100%; Dog: 100%; Goat: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

The protein encoded by this gene belongs to the derlin family, and resides in the endoplasmic reticulum (ER). Proteins that are unfolded or misfolded in the ER must be refolded or degraded to maintain the homeostasis of the ER. This protein appears to be involved in the degradation of misfolded glycoproteins in the ER. Several alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DERL3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DERL3
Gene Accession No. Q96Q80
Gene Alias 1810006I20Rik; 1810063P04Rik; C22orf14; Degradation in endoplasmic reticulum protein 3; Der1-like domain family, member 3; der1-like protein 3; Der3; Derl3; derlin 3; derlin-3; DERtrin 3; DERtrin-3; hypothetical protein MGC63991; IZP6; LLN2; MGC71803; Protein IZP6; Unknown (protein for MGC:142482); zgc:63991
Gene Symbols DERL3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DERL3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91319
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.