Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Derlin 1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB101754 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB101754 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB101754 Supplier Novus Biologicals Supplier No. NBP297369B
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Derlin 1 Polyclonal specifically detects Derlin 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Derlin 1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias DER-1, DER1Degradation in endoplasmic reticulum protein 1, Der1-like domain family, member 1, Der1-like protein 1, derlin-1, DERtrin-1, FLJ13784, FLJ42092, MGC3067, PRO2577
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Derlin 1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ER Markers, Immunology, Lipid and Metabolism, Protein Turnover, Signal Transduction, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 79139
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.