Description
Includes
TACS BlueLabel, Red Counterstain C, and reagents necessary for generating positive controls
Incorporation of brominated deoxyuridine (BrdU), which is more efficiently incorporated by TdT than biotinylated nucleotides at DNA fragmentation sites, is detected using a highly specific and sensitive biotinylated anti-BrdU antibody in combination with
Specifications
Specifications
| Quantity | 30 Tests |
| Content And Storage | The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| Product Line | R&D Systems |