Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dermatopontin Mouse anti-Human, Rat, Clone: 4B1A12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Dermatopontin |
---|---|
Clone | 4B1A12 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6779
![]() ![]() |
Proteintech Group Inc
663481IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin.Specifications
Dermatopontin | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
dermatopontin, DPT, TRAMP | |
Dpt | |
IgG2b | |
Protein A | |
Antibody |
4B1A12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
Q07507 | |
1805, 289178 | |
Dermatopontin Fusion Protein Ag6200 | |
Primary | |
-20°C | |
DPT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title