Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Deschloroclozapine
GREENER_CHOICESDP

Catalog No. 719310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-931-0 10 mg
71-935-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 71-931-0 Supplier Tocris Bioscience™ Supplier No. 7193/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity and highly potent activator of hM3Dq and hM4Di DREADDs; rapidly blood brain barrier penetrant

Deschloroclozapine is a high affinity and highly potent activator of muscarinic DREADDs (Ki values are 6.3 nM for hM3Dq and 4.2 nM for hM4Di, in vitro; EC50 values are 0.13 nM for hM3Dq and 0.081 nM for hM4Di, in vitro). Derivative of clozapine (Cat. No. 0444). Exhibits 100-fold greater affinity for hM3Dq and hM4Di compared to clozapine N-oxide (CNO; Cat. No. 4936), and 50-fold greater affinity compared to DREADD agonist 21 (Cat. No. 5548). Displays low 'off-target' receptor binding (Ki >100 nM at majority of GPCRs, ion channels and transporters tested). Displays rapid (approx. 10 minutes after i.p injection) binding and activation of hM3Dq and hM4Di in mice and non-human primates. Induces spatial memory deficits in non-human primates expressing hM4Di in the prefrontal cortex.

Specifications

Chemical Name or Material 11-(4-Methyl-1-piperazinyl)-5 H-dibenzo[b,e][1,4 ]diazepine
CAS 7-7-1977
Quantity 10 mg
Target DREADD Activators
Molecular Formula C18H20N4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.