Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DESI1 Polyclonal Antibody
DESI1 Polyclonal Antibody
Supplier: Thermo Scientific PA571343
Description
DESI1 Polyclonal Antibody for Western Blot

Specifications
DESI1 | |
Polyclonal | |
Unconjugated | |
DESI1 | |
D15Wsu75e; DESI1; DeSI-1; DESI2; desumoylating isopeptidase 1; desumoylating isopeptidase 2; DJ347H13.4; FAM152B; family with sequence similarity 152, member B; Polyubiquitinated substrate transporter; POST; PPPDE peptidase domain containing 2; PPPDE peptidase domain-containing protein 2; PPPDE2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27351 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6ICB0 | |
DESI1 | |
synthetic peptide directed towards the C-terminal region of human DESI1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction