Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmin Antibody (CL4501), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Desmin |
---|---|
Dilution | Western Blot 1 μg/mL, Immunocytochemistry/Immunofluorescence 2-10 μg/mL |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Desmin Monoclonal specifically detects Desmin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Desmin | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cytoskeleton Markers | |
CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
DES | |
IgG1 | |
Protein A purified |
Western Blot 1 μg/mL, Immunocytochemistry/Immunofluorescence 2-10 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
1674 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:HTKKTVMIKTIETRDGE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title