Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmin Antibody (DES/1711) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$648.50
Specifications
Antigen | Desmin |
---|---|
Clone | DES/1711 |
Concentration | 1 mg/mL |
Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254503100
![]() |
Novus Biologicals
NBP254503100UG |
100 μg |
Each for $648.50
|
|
|||||
NB254503200
![]() |
Novus Biologicals
NBP254503200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Desmin Monoclonal specifically detects Desmin in Human, Mouse, Rat, Chicken, Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
Desmin | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Cytoskeleton Markers | |
PBS with No Preservative | |
1674 | |
Recombinant human full-length desmin protein | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
DES/1711 | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Chicken, Hamster | |
CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
DES | |
IgG1 κ | |
Protein A or G purified | |
52 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title