Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Desmin Antibody (DES/3255), Alexa Fluor™ 647, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314044AF647
                        
                            
                        
                        
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
Desmin Monoclonal antibody specifically detects Desmin in Human, Bovine, Canine, Equine, Feline samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Desmin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| Recombinant full-length human desmin protein (Uniprot: P17661 ) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 1674 | |
| Store at 4°C in the dark. | |
| IgG2b κ | 
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DES/3255 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Canine, Equine, Feline | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction