Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmin Antibody (DES/3255), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314044R
This item is not returnable.
View return policy
Description
Desmin Monoclonal antibody specifically detects Desmin in Human, Bovine, Canine, Equine, Feline samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Desmin | |
| Monoclonal | |
| DyLight 550 | |
| CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| Recombinant full-length human desmin protein (Uniprot: P17661 ) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 1674 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DES/3255 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Canine, Equine, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction