Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmin (DES) Monoclonal Antibody (OTI3B12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF814318


Description
Desmin (DES) Monoclonal antibody specifically detects Desmin (DES) in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Desmin (DES) | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P07196 | |
Mouse | |
Protein A/G | |
RUO | |
4747 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin) | |
OTI3B12 | |
Unconjugated | |
NEFL | |
CMD1I; CSM1; CSM2; DES; Desmin; desmin-like protein; FLJ12025; FLJ39719; FLJ41013; FLJ41793; I79_018932; intermediate filament protein; LGMD2R; MFM1; muscle-specific intermediate filament desmin; muscle-specific intermediate filament protein; mutant desmin p.K241E; SCPNK; similar to desmin | |
Human recombinant protein fragment corresponding to amino acids 401-543aa of human NEFL (NP_006149) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction