Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmin Mouse anti-Human, Bovine, Canine, Equine, Feline, Clone: DES/3255, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313866100UG
This item is not returnable.
View return policy
Description
Desmin Monoclonal antibody specifically detects Desmin in Human, Bovine, Canine, Equine, Feline samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Desmin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
P17661 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Bovine, Canine, Equine, Feline | |
Purified |
Immunohistochemistry (Paraffin) | |
DES/3255 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
Recombinant full-length human desmin protein (Uniprot: P17661 ) | |
100 μg | |
Cell Biology, Cellular Markers | |
1674 | |
Store at 4°C. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction