Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmin Antibody (RD301), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197707R
Description
Desmin Monoclonal specifically detects Desmin in Human, Mouse, Rat, Porcine, Canine, Chicken, Hamster, Rabbit, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Desmin | |
Monoclonal | |
DyLight 550 | |
50 mM sodium borate with 0.05% sodium azide | |
DES | |
Clone RD301 is a mouse monoclonal IgG2b antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a mouse immunized with a cytoskeletal desmin extract of chicken gizzard. | |
Primary | |
RD301 reacts exclusively with desmin, which is expressed in smooth and striated muscle cells and their tumors e.g. rhabdomyosarcoma and leiomyosarcoma. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
RD301 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
Mouse | |
0.1 mL | |
1674 | |
Human, Mouse, Rat, Pig, Canine, Chicken, Hamster, Rabbit, Zebrafish | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction