Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Mouse anti-Human, Rat, Mouse, Clone: 4C12.1, MilliporeSigma™

Catalog No. MABT535MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT535MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT535MI Supplier MilliporeSigma Supplier No. MABT535
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Desmin Antibody, clone 4C12.1 is a mouse monoclonal antibody that targets Desmin in Human, Mouse, and Rat and it is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4C12.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols DES
Host Species Mouse
Immunogen GST/His-taged recombinant fragment corresponding to 64 amino acids from the C-terminal region of human Desmin.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001918
Test Specificity Clone 4C12.1 specifically detects Desmin in human, mouse, and rat muscles. It targets an epitope within 64 amino acids from the C-terminal end.
Target Species Human, Rat, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.