Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Desmin (Muscle Cell Marker) Ab-1, Mouse Monoclonal Antibody

Catalog No. MS376S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS376S 1 mL Concentrated
MS376S0 100 μL Concentrated
MS376S1 500 μL Concentrated
MS376R7 7 mL Prediluted
MS376RQ 12 mL Ready-to-Use
MS376R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS376S Supplier Epredia™ Supplier No. MS376S
Add to Cart
Add to Cart

Specifically detect desmin in human, baboon, monkey, cow, cat, dog, hamster, rat and chicken samples.

Desmin is an intermediate filament protein of both smooth and striated muscles. Antibody to desmin reacts with striated (skeletal and cardiac) as well as smooth muscle cells. In skeletal and cardiac muscles, the staining is confined to the Z-bands giving a characteristic striated appearance. Anti-desmin antibody is useful in identification of tumors of myogenic origin. It reacts with leiomyosarcomas (smooth muscle) as well as rhabdomyosarcomas (striated muscle).

Host Species: Mouse

Clone: D33

Isotype: IgG1/κ

Species Reactivity: Human, Baboon, Monkey, Cow, Cat, Dog, Hamster, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Desmin from human muscle

Molecular Weight: 53 kDa

Positive Control: Muscle or Sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Antigen Desmin (Muscle Cell Marker) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone D33
Conjugate Unconjugated
Host Species Mouse
Immunogen Desmin from human muscle
Quantity 1 mL
Regulatory Status IVD
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Feline, Hamster, Human, Non-human Primate, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.