Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmocollin-1 Antibody (772906) [DyLight 405], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7367E
Description
Desmocollin-1 Monoclonal antibody specifically detects Desmocollin-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOFSpecifications
Desmocollin-1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1823 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
772906 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, desmosomal glycoprotein 2/3, DG2/DG3 | |
Mouse myeloma cell line NS0-derived recombinant mouse Desmocollin-1, Arg135-Lys691, Accession # P55849 | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction