Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmocollin-2 Antibody (003), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290904APC
Description
Desmocollin-2 Monoclonal antibody specifically detects Desmocollin-2 in Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Desmocollin-2 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
1824 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
003 | |
Immunohistochemistry-Paraffin | |
ARVD11, Cadherin family member 2, CDHF2DGII/III, desmocollin 2, desmocollin-2, Desmocollin-3, Desmosomal glycoprotein II, desmosomal glycoprotein II/III, Desmosomal glycoprotein III, DG2, DSC3DKFZp686I11137 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat Desmocollin-2 (Uniprot#: Q3T1K6; Met1-Pro694). | |
0.1 mL | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction