Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Desmocollin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582465
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582465 Supplier Invitrogen™ Supplier No. PA582465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KTVIICKPTM SSAEIVAVDP DEPIHGPPFD FSLESSTSEV QRMWRLKAIN DTAARLSYQN DPPFGSYVVP ITVRDRLGMS SVTSLDVTLC DCITENDCTH RVDPRIGGGG VQLGKW Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 74%.

This gene encodes a member of the desmocollin protein subfamily. Desmocollins, along with desmogleins, are cadherin-like transmembrane glycoproteins that are major components of the desmosome. Desmosomes are cell-cell junctions that help resist shearing forces and are found in high concentrations in cells subject to mechanical stress. This gene is found in a cluster with other desmocollin family members on chromosome 18. Mutations in this gene are associated with arrhythmogenic right ventricular dysplasia-11, and reduced protein expression has been described in several types of cancer. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmocollin 2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DSC2
Gene Accession No. Q02487
Gene Alias ARVC10; ARVD10; ARVD11; AW228162; Cadherin family member 2; CDHF2; CDHF5; CDHF6; CMD1BB; desmocollin 2; desmocollin-2; Desmocollin-3; desmoglein-2; desmoglein-3; Desmosomal glycoprotein II; desmosomal glycoprotein II/III; Desmosomal glycoprotein III; DG2; DGII/III; DSC2; Dsc2a; Dsc2b; DSC3; Epithelial type 2 desmocollin; HDGC; PVA
Gene Symbols DSC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DSC2. Recombinant protein control fragment (Product #RP-90654).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1824
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.