Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Desmocollin 2 Recombinant Rabbit Monoclonal Antibody (004)
GREENER_CHOICE

Catalog No. PIMA530496
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530496 Supplier Invitrogen™ Supplier No. MA530496
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human DSC2.

This gene encodes a member of the desmocollin protein subfamily. Desmocollins, along with desmogleins, are cadherin-like transmembrane glycoproteins that are major components of the desmosome. Desmosomes are cell-cell junctions that help resist shearing forces and are found in high concentrations in cells subject to mechanical stress. This gene is found in a cluster with other desmocollin family members on chromosome 18. Mutations in this gene are associated with arrhythmogenic right ventricular dysplasia-11, and reduced protein expression has been described in several types of cancer. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmocollin 2
Applications ELISA
Classification Recombinant Monoclonal
Clone 004
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DSC2
Gene Accession No. Q02487
Gene Alias ARVC10; ARVD10; ARVD11; AW228162; Cadherin family member 2; CDHF2; CDHF5; CDHF6; CMD1BB; desmocollin 2; desmocollin-2; Desmocollin-3; desmoglein-2; desmoglein-3; Desmosomal glycoprotein II; desmosomal glycoprotein II/III; Desmosomal glycoprotein III; DG2; DGII/III; DSC2; Dsc2a; Dsc2b; DSC3; Epithelial type 2 desmocollin; HDGC; PVA
Gene Symbols DSC2
Host Species Rabbit
Immunogen Recombinant Human DSC2 Protein (Met1-Arg684).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1824
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.