Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Desmocollin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547909
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547909 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547909 Supplier Invitrogen™ Supplier No. PA547909
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 25% cross-reactivity with recombinant human Desmocollin-3 is observed, and approximately 5% cross-reactivity with recombinant mouse (rm) Desmocollin-1 and rmDesmocollin-2 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmocollin 3
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Dsc3
Gene Accession No. P55850
Gene Alias 5430426I24Rik; ARVC10; ARVD10; Cadherin family member 3; CDHF3; CDHF5; CDHF6; CMD1BB; desmocollin 3; desmocollin-3; Desmocollin-4; desmoglein-2; desmoglein-3; DSC; DSC1; DSC2; DSC3; DSC4; HDGC; HT-CP; PVA
Gene Symbols Dsc3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Desmocollin-3 Arg136-Leu695.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13507
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.