Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmocollin-3 Rabbit anti-Human, Clone: 7H1O6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Desmocollin-3 |
---|---|
Clone | 7H1O6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Desmocollin-3 Monoclonal antibody specifically detects Desmocollin-3 in Human samples. It is validated for Western BlotSpecifications
Desmocollin-3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Cadherin family member 3, CDHF3HT-CP, desmocollin 3, desmocollin-4, DSC, DSC1, DSC2, DSC4desmocollin-3 | |
A synthetic peptide corresponding to a sequence within amino acids 797-896 of human Desmocollin-3 (Q14574). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
7H1O6 | |
Monoclonal | |
Purified | |
RUO | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1825 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title