Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Desmocollin 3 Recombinant Rabbit Monoclonal Antibody (7H1O6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542595
This item is not returnable.
View return policy
Description
Positive test controls include: TE-1. Immunogen sequence: HHHTLDSCRG GHTEVDNCRY TYSEWHSFTQ PRLGEKLHRC NQNEDRMPSQ DYVLTYNYEG RGSPAGSVGC CSEKQEEDGL DFLNNLEPKF ITLAEACTKR.
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
Specifications
Desmocollin 3 | |
Recombinant Monoclonal | |
0.4 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14574 | |
Dsc3 | |
A synthetic peptide corresponding to a sequence within amino acids 797-896 of human Desmocollin 3 (Q14574). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
7H1O6 | |
Unconjugated | |
Dsc3 | |
5430426I24Rik; ARVC10; ARVD10; Cadherin family member 3; CDHF3; CDHF5; CDHF6; CMD1BB; desmocollin 3; desmocollin-3; Desmocollin-4; desmoglein-2; desmoglein-3; DSC; DSC1; DSC2; DSC3; DSC4; HDGC; HT-CP; PVA | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1825 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction