Learn More
Invitrogen™ Desmocollin 3 Recombinant Rabbit Monoclonal Antibody (JE55-86)

Description
Positive Control: Rat skin tissue lysate, A431 cell lysate, 293T, human skin tissue, mouse esophagus tissue, A431. Subcellular Location: Cell membrane, desmosome.
Specifications
Specifications
| Antigen | Desmocollin 3 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | JE55-86 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Dsc3 |
| Gene Accession No. | P55850, Q14574 |
| Gene Alias | 5430426I24Rik; ARVC10; ARVD10; Cadherin family member 3; CDHF3; CDHF5; CDHF6; CMD1BB; desmocollin 3; desmocollin-3; Desmocollin-4; desmoglein-2; desmoglein-3; DSC; DSC1; DSC2; DSC3; DSC4; HDGC; HT-CP; PVA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.