Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoglein-1,2 Antibody (33-3D), Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Desmoglein-1,2 |
---|---|
Clone | 33-3D |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Electron Microscopy |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Electron Microscopy |
Description
Desmoglein-1,2 Monoclonal antibody specifically detects Desmoglein-1,2 in Human, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Electron MicroscopySpecifications
Desmoglein-1,2 | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunofluorescence, Electron Microscopy | |
Unconjugated | |
Mouse | |
Cancer, Cellular Signaling | |
PBS | |
1828 | |
IgM | |
Thiophilic Chromatography |
33-3D | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Electron Microscopy | |
Monoclonal | |
Purified | |
RUO | |
Human, Bovine | |
Cadherin family member 4, Cadherin family member 5, desmoglein 1, desmoglein 2, desmoglein-1, desmoglein-2, Desmosomal glycoprotein 1, DG1 | |
Bovine Desmoglein-1 from nasal epidermis | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title