Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Desmoglein 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121240 Supplier Invitrogen™ Supplier No. PA5121240
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A-431, HeLa, 293T, BT-474. The target is usually found in the following locations: Cell junction, Cell membrane, Single-pass type I membrane protein, desmosome. Immunogen sequence: SPHFGTTTVI SESTYPSGPG VLHPKPILDP LGYGNVTVTE SYTTSDTLKP SVHVHDNRPA SNVVVTERVV GPISGADLHG MLEMPDLRDG SNVIVTERVI APSSSLPTSL TIHHPRESSN VVVTERVIQP TSGMIGSLSM HPELANAHNV IVTERVVSGA GVTGISGTTG ISGGIGSSGL VGTSMGAGSG ALSGAGISGG GIGLSSLGGT ASIGHMRSSS DHHFNQTIGS ASPSTARSRI TKYSTVQYSK.

Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoglein 1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DSG1
Gene Accession No. Q02413
Gene Alias Cadherin family member 4; CDHF4; desmoglein 1; desmoglein 1 beta; desmoglein 1 gamma; desmoglein-1; Desmosomal glycoprotein 1; DG1; DSG; DSG1; Dsg1b; Dsg1c; EPKHE; EPKHIA; Pemphigus foliaceus antigen; PPKS1; SPPK1
Gene Symbols DSG1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 800-1049 of human DSG1 (NP_001933.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1828, 291755
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.