Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoglein-2 Antibody (22H6), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Desmoglein-2 |
---|---|
Clone | 22H6 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
Desmoglein-2 Monoclonal antibody specifically detects Desmoglein-2 in Human samples. It is validated for ELISASpecifications
Desmoglein-2 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ARVC10, ARVD10, Cadherin family member 5, CDHF5MGC117034, CMD1BB, desmoglein 2, desmoglein-2, HDGC, MGC117036, MGC117037 | |
Recombinant Human Desmoglein-2 protein [UniProt Q14126] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
22H6 | |
ELISA | |
Unconjugated | |
Human | |
Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
1829 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title