Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoglein-2 Antibody (6D8) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$646.00

Specifications

Antigen Desmoglein-2
Clone 6D8
Concentration 1 mg/mL
Dilution Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254530100
SDP
View Documents
Novus Biologicals
NBP254530100UG
100 μg
Each for $646.00
Only null left
Add to Cart
 
NB254530200
SDP
View Documents
Novus Biologicals
NBP254530200UG
200 μg N/A N/A N/A
Description

Description

Desmoglein-2 Monoclonal specifically detects Desmoglein-2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications

Specifications

Desmoglein-2
1 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
Unconjugated
Mouse
Human
ARVC10, ARVD10, Cadherin family member 5, CDHF5MGC117034, CMD1BB, desmoglein 2, desmoglein-2, HDGC, MGC117036, MGC117037
DSG2
IgG1 κ
Protein A or G purified
165 kDa
6D8
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
PBS with No Preservative
1829
Recombinant human full-length DSG2 protein
Primary
Store at -20 to -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.