Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoglein-2 Antibody ([8E5), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254529AF594
Description
Desmoglein-2 Monoclonal specifically detects Desmoglein-2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Desmoglein-2 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
DSG2 | |
Recombinant human Desmoglein-2 protein (Uniprot: Q14126) | |
Primary | |
Recognizes a protein of 165kDa, identified as Desmoglein-2 (DSG2). This monoclonal antibody recognizes the extracellular domain of human desmoglein-2. Desmoglein-2 is a member of the desmosomal cadherin family. Desmosomes are intercellular adhering junctions that represent cell surface attachment sites for intermediate filament. Desmocollins and desmogleins are the main desmosomal transmembrane proteins. Desmogleins consist of Dsg1, Dsg2, Dsg3, and Dsg4 isoforms. Within the desmosome, the extracellular domain of desmoglein is essential for calcium dependent heterophilic binding to the desmocollins, whereas the intracellular domain is essential for binding to the desmosomal plaque protein, plakoglobin. Human Desmoglein-2 is a type I transmembrane glycoprotein of 1117 amino acid (aa) residues with a 23 aa signal peptide and a 25 aa propeptide. It differs from other classic cadherins by having four instead of five cadherin repeat domains in its extracellular region, and a much larger cytoplasmic region containing five desmoglein repeat domains which share homology with the cadherin repeats. Instead of having the HAV adhesion motif found in type I cadherins, desmogleins have R/YAL as the adhesion motif on its amino-terminal cadherin repeat. The cytoplasmic tails of desmogleins interact with desmoplakins, plakoglobin and plakophilins. In turn, these proteins link the desmogleins with the intermediate filaments. Desmoglein-2 has been shown to be important in establishing cell-cell adhesion and function in epithelial cells. Desmoglein2 was originally identified in colon carcinoma and colon, and was named HDGC (human desmoglein colon). | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
[8E5] | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
ARVC10, ARVD10, Cadherin family member 5, CDHF5MGC117034, CMD1BB, desmoglein 2, desmoglein-2, HDGC, MGC117036, MGC117037 | |
Mouse | |
0.1 mL | |
1829 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction