Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoglein 2 Monoclonal Antibody (3G132), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA191589
Description
MA1-91589 detects Desmoglein 2 from human samples. MA1-91589 has been successfully used in immunofluorescence, immunoprecipitation, Western blot applications. The MA1-91589 immunogen is: Full length native protein (purified) (Human).
The product of this gene is part of a large complex localized to the cytoplasm, nucleoli, and to discrete nuclear bodies called Gemini bodies. The complex functions in spliceosomal snRNP assembly in the cytoplasm, and regenerates spliceosomes required for pre-mRNA splicing in the nucleus. The encoded protein directly interacts with a DEAD box protein and several spliceosome core proteins. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
Desmoglein 2 | |
Monoclonal | |
0.5 mg/mL | |
DSG2 | |
ARVC10, ARVD10, CDHF5, CMD1BB, HDGC, MGC117034, MGC117036, MGC117037 | |
Mouse | |
Protein A | |
RUO | |
1829 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
3G132 | |
Unconjugated | |
Q14126 | |
DSG2 | |
Full length native protein (purified) (Human). | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction