Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Desmoglein 2 Monoclonal Antibody (CSTEM28), eBioscience™, Invitrogen™

Catalog No. 14915937
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-915-937 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-915-937 Supplier Invitrogen™ Supplier No. 14915937
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This MAb CSTEM28 reacts with human desmoglein 2 (DSG2), a type 1 transmembrane protein belonging to the cadherin family. DSG2 is expressed by epithelial cells, cardiomyocytes, endothelial progenitors, and CD34+CD45dim hematopoietic stem and progenitor cells. This adhesion molecule forms cell-cell adhesion complexes called desmosomes that provide tensile strength to tissues that experience mechanical stress. Other than its role in desmosome formation, DSG2 plays a role in barrier integrity, adhesion, migration, cell apoptosis, proliferation, vasculogenic mimicry, and intracellular signaling. DSG2-expressing endothelial progenitor cells are pro-angiogenic, suggesting a role in vasculature formation. On hematopoietic stem and progenitor cells, DSG2 expression is highest on CD38-CD90+ multi-potent cells. DSG2 expression is progressively decreased as a function of differentiation, with DSG2 expression absent on mature lymphocytes. DSG2 also serves as a receptor for adenoviruses and can be shed by mucosal barriers in the intestine.

Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoglein 2
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone CSTEM28
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene DSG2
Gene Accession No. Q14126
Gene Alias AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; DSG2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037
Gene Symbols DSG2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1829
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.