Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoglein 2 Monoclonal Antibody (ZY50), Abbomax

Catalog No. 502204386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4386 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4386 Supplier Abbomax Inc Supplier No. 605770
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Human thyroid gland papillary carcinoma Cellular location: Cell membrane, Cell junction.

Desmoglein 2 Monoclonal antibody specifically detects Desmoglein 2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Desmoglein 2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZY50
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene DSG2
Gene Accession No. Q14126
Gene Alias AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; DSG2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037
Host Species Mouse
Immunogen A partial recombinant protein of human Desmoglein 2
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1829
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.