Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoglein 2 Mouse anti-Human, PE, Clone: 6D8 (also known as 6D8), BD Pharmingen™
Mouse Monoclonal Antibody
$154.70 - $384.00
Specifications
Antigen | Desmoglein 2 |
---|---|
Clone | 6D8 (also known as 6D8) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
Desmoglein 2 Monoclonal antibody specifically detects Desmoglein 2 in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
Desmoglein 2 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
DSG2; CDHF5; Cadherin family member 5; Desmoglein-2; desmoglein-2 | |
IgG1 κ | |
Affinity Chromatography |
6D8 (also known as 6D8) | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Full-length recomb. Desmoglein 2 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title