Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoglein-3 Antibody (5G11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications
$206.50 - $499.50
Specifications
Antigen | Desmoglein-3 |
---|---|
Clone | 5G11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:1000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Dot Blot reported in scientific literature (PMID 15588331) |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1001643SS
![]() |
Novus Biologicals
NB1001643SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB1001643
![]() |
Novus Biologicals
NB1001643 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Desmoglein-3 Monoclonal specifically detects Desmoglein-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot.Specifications
Desmoglein-3 | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cytoskeleton Markers, Extracellular Matrix, Signal Transduction | |
P32926 | |
1830 | |
Human Desmoglein 3 recombinant protein [UniProt# P32926] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5G11 | |
Western Blot 1:500 - 1:1000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Dot Blot reported in scientific literature (PMID 15588331) | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
DSG3 | |
IgG1 | |
Protein G purified | |
This antibody reacts with Desmoglein-3. Does not cross-react with Desmoglein 1 or Desmoglein 2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title