Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoglein-3 Antibody (5G11), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1001643G
Description
Desmoglein-3 Monoclonal specifically detects Desmoglein-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Desmoglein-3 | |
Monoclonal | |
DyLight 488 | |
P32926 | |
DSG3 | |
Human Desmoglein 3 recombinant protein [UniProt# P32926] | |
0.1 mL | |
Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction | |
1830 | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
5G11 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This antibody reacts with Desmoglein-3. Does not cross-react with Desmoglein 1 or Desmoglein 2. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction