Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoglein-3 Antibody (5H10), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP178984AF700
Description
Desmoglein-3 Monoclonal specifically detects Desmoglein-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Desmoglein-3 | |
Monoclonal | |
Alexa Fluor 700 | |
P32926 | |
DSG3 | |
The extracellular domain of human Desmoglein 3 [Swiss-Prot# P32926] | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
5H10 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
Mouse | |
130 kDa | |
0.1 mL | |
Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction | |
1830 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction