Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoglein-3 Antibody (DSG3/2840), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308509AF488
Description
Desmoglein-3 Monoclonal antibody specifically detects Desmoglein-3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
Desmoglein-3 | |
Monoclonal | |
Alexa Fluor 488 | |
Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
Recombinant fragment (around aa 379-491) human Desmoglein-3 protein (exact sequence is proprietary) (Uniprot: P32926) | |
100 μg | |
Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction | |
1830 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin), Protein Array | |
DSG3/2840 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction