Learn More
Invitrogen™ Desmoglein 3 Monoclonal Antibody (5G11)

Description
Desmoglein 3 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow, IP
Specifications
Specifications
| Antigen | Desmoglein 3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5G11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Dsg3 |
| Gene Accession No. | P32926 |
| Gene Alias | 130 kDa pemphigus vulgaris antigen; 130 kDa pemphigus vulgaris antigen homolog; bal; balding; Cadherin family member 6; CDHF6; desmoglein 3; desmoglein-3; DKFZp686P23184; Dsg3; PVA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.