Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Desmoglein 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584316 Supplier Invitrogen™ Supplier No. PA584316
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FRPASKTFTV QKGISSKKLV DYILGTYQAI DEDTNKAASN VKYVMGRNDG GYLMIDSKTA EIKFVKNMNR DSTFIVNKTI TAEVLAIDEY TGKTSTGTVY VRVPDFND Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 80%.

Desmoglein 3 (Dsg3), also known as Cadherin family member 6 (CDHF6), is a member of the desmosomal cadherin family and plays a critical role in cell-cell adhesion. It is predominately expressed in stratified squamous epithelium, including the skin, oral mucosa, and esophagus. Desmoglein 3 has been characterized as the auto-antigen of pemphigus vulgaris (PV), a life-threatening autoimmune disease. Binding of anti-Desmoglein 3 autoantibodies causes the loss of cell-cell adhesion, which clinically manifests as blisters on the skin and oral mucosa, and histologically as suprabasilar acantholysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoglein 3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dsg3
Gene Accession No. P32926
Gene Alias 130 kDa pemphigus vulgaris antigen; 130 kDa pemphigus vulgaris antigen homolog; bal; balding; Cadherin family member 6; CDHF6; desmoglein 3; desmoglein-3; DKFZp686P23184; Dsg3; PVA
Gene Symbols Dsg3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DSG3. Recombinant protein control fragment (Product #a href=.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1830
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.