Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Desmoglein 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537155 Supplier Invitrogen™ Supplier No. PA537155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 114 kDa. Purity is >95% by SDS-PAGE.

Desmoglein proteins are cadherin-type cell adhesion molecules. Desmoglein 4 (dsg4) shares 41% identity with human desmoglein 1, 37% with human desmoglein 2 and 50% with human desmoglein 3. A type I membrane protein of the cadherin protein family, dsg4 is expressed in salivary gland, suprabasal epidermis, hair follicle, testis, prostate and skin. In the hair follicle, dsg4 is an important mediator of keratinocyte cell adhesion and coordinates the transition from proliferation to differentiation. The human DSG4 gene is composed of 16 exons spanning approximately 37 kb of 18q12 and is situated between DSG1 and DSG3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoglein 4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene DSG4
Gene Accession No. Q6W3B0, Q7TMD7, Q86SJ6
Gene Alias Cadherin family member 13; CDGF13; CDHF13; desmoglein 4; Desmoglein4; desmoglein-4; DSG4; HYPT6; LAH; lanceolate hair
Gene Symbols DSG4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in human Desmoglein 4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147409, 16769, 291754
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.