Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desmoplakin Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP149879
Description
Desmoplakin Polyclonal specifically detects Desmoplakin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Desmoplakin | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
250/210 kDa paraneoplastic pemphigus antigen, desmoplakin, desmoplakin (DPI, DPII), desmoplakin I, desmoplakin II, DP, DPI, DPII, KPPS2, PPKS2 | |
Goat | |
Affinity Purified | |
RUO | |
1832 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 3 - 6 ug/ml, Immunohistochemistry-Paraffin 3 - 6 ug/ml, Peptide ELISA Detection limit 1:4000 | |
NP_004406.2 | |
DSP | |
Peptide with sequence C-QESVKQAHLRNEH corresponding to internal region according to NP_004406.2. | |
0.1 mg | |
Primary | |
This antibody is expected to recognize reported isoform I (NP_004406.2) only. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction