Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmoplakin, Mouse anti-Human, Clone: 20B6, Millipore Sigma™

Catalog No. MABT149225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT149225 25 μg
MABT1492100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT149225 Supplier MilliporeSigma Supplier No. MABT149225UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Desmoplakin, clone 20B6, Cat. No. MABT1492, is a mouse monoclonal antibody that detects human Desmoplakin and has been tested for use in Immunocytochemistry and Western Blotting.

Desmoplakin (UniProt: P15924; also known as DP, 250/210 kDa paraneoplastic pemphigus antigen) is encoded by the DSP gene (Gene ID:1832) in human. Desmoplakin is a major high molecular weight homodimeric protein that is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes, which are prominent adhesive junctions present between many epithelial cells and cardiomyocytes. Desmoplakin is the most abundant of the desmosomal proteins and plays a key role in tissue morphogenesis and maintaining the integrity of tissue. Desmoplakin is composed of an N-terminal plakin domain, a central alpha-helical coiled-coil rod domain, and a C-terminal intermediate filament-binding domain. The N-terminal domain is required for desmosome association and the C-terminal binds to desmin. Three isoforms for Desmoplakin have been reported that are produced by alternative splicing. Desmoplakin 2 and 3 lack most of the central rod domain. Desmoplakin 1 is the dominant isoform in the cardiac tissue. Desmoplakin is phosphorylated by protein kinase A on serine 2849 and this phosphorylation affects its association with epidermal, simple cytokeratins, and VIM intermediate filaments. Mutations in DSP gene have been linked to DCWHK (Cardiomyopathy, dilated, with woolly hair and keratoderma) that is characterized by a generalized striate keratoderma particularly affecting the palmoplantar epidermis, woolly hair, and dilated left ventricular cardiomyopathy. Mice deficient in Desmoplakin perish around the time of implantation (E6.5) and show fewer desmosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoplakin
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 20B6
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected Desmoplakin in Western Blotting applications (Sobolik-Delmaire, T., et. al. (2006). J Biol Chem. 281(25):16962-70; Hall, C., et. al. (2009). Cell Commun Adhes. 16(1-3):15-27; Roberts, B.J., et. al. (2011). Exp Cell Res. 317(20):2814-22).Immunocytochemistry Analysis: A representative lot detected Desmoplakin in Immunocytochemistry applications (Roberts, B.J., et. al. (2011). Exp Cell Res. 317(20):2814-22; Sobolik-Delmaire, T., et. al. (2006). J Biol Chem. 281(25):16962-70; Roberts, B.J., et. al. (2016). J Biol Chem. 291(48):24857-24865).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias DP;250/210 kDa paraneoplastic pemphigus antigen
Gene Symbols DSP
Host Species Mouse
Immunogen His-tagged recombinant fragment corresponding to 863 amino acids from the C-terminal half of human Desmoplakin.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001008844
Test Specificity Clone 20B6 detects human Desmoplankin. It targets an epitope with in the C-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.