Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Desmoplakin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561103 Supplier Invitrogen™ Supplier No. PA561103
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NHNKVIETNR ENDKQETWML MELQKIRRQI EHCEGRMTLK NLPLADQGSS HHITVKINEL KSVQNDSQAI AEVLNQLKDM LANFRGSEKY CYLQNEVF Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoplakin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DSP
Gene Accession No. P15924
Gene Alias 2300002E22Rik; 250/210 kDa paraneoplastic pemphigus antigen; 5730453H04Rik; AA407887; AA407888; AW109828; DCWHKTA; dermatan sulfate proteoglycan 3; desmoplakin; desmoplakin I/II; Desmoplakin II; desmoplakin; LOW QUALITY PROTEIN: desmoplakin; Desmoplakin-I; desmosomal cytoskeletal connector molecule; DP; Dsp; dspg3; Oculoglycan; Opt; Optc; optcl; Opticin; rul; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047
Gene Symbols DSP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Desmoplakin. Recombinant protein control fragment (Product #RP-104081).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.