Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dess-Martin Periodinane 95.0+%, TCI America™
SDP

Catalog No. D204525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D204525G 25 g
D20451G 1 g
D20455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D204525G Supplier TCI America Supplier No. D204525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 87413-09-0
Molecular Formula C13H13IO8
Molecular Weight (g/mol) 424.14
MDL Number MFCD00130127
InChI Key NKLCNNUWBJBICK-UHFFFAOYSA-N
Synonym dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane
PubChem CID 159087
IUPAC Name 1,1-bis(acetyloxy)-3-oxo-3H-1λ⁵,2-benziodaoxol-1-yl acetate
SMILES CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 25 g
UN Number 1479
Formula Weight 424.14
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dess-Martin Periodinane
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.