Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Destrin 19 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987388
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7388 Supplier Bioss Supplier No. BS6630R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene.

Specifications

Antigen Destrin 19
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DSTN
Gene Accession No. P60981
Gene Alias 2610043P17Rik; ACTDP; Actin-depolymerizing factor; ADF; AU042046; bA462D18.2; bA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)); corn1; corneal disease 1; Destrin; destrin (actin depolymerizing factor); destrin, actin depolymerizing factor; DSN; Dstn; epididymis luminal protein 32; HEL32; RP23-77A2.1; sID; Sid 23; Sid23; sid23p
Gene Symbols DSTN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Destrin 19/ADF.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11034
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.