Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Destrin Antibody (OTI2F7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271669B
Description
Destrin Monoclonal specifically detects Destrin in Human, Mouse, Canine, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Destrin | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
DSTN | |
Full length recombinant protein of human Destrin (NP_006861) produced in HEK293T cells. | |
0.1 mL | |
11034.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2F7 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ACTDPdestrin, Actin-depolymerizing factor, ADFbA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)), bA462D18.2, destrin (actin depolymerizing factor), DSN | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Canine, Monkey, Primate | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction