Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Destrin Antibody (OTI2F7), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271669MFV500
Description
Destrin Monoclonal antibody specifically detects Destrin in Human, Mouse, Canine, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
Destrin | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Canine, Primate, Monkey | |
Purified |
Flow Cytometry | |
OTI2F7 | |
Flow Cytometry | |
ACTDPdestrin, Actin-depolymerizing factor, ADFbA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)), bA462D18.2, destrin (actin depolymerizing factor), DSN | |
Full length human recombinant protein of human DSTN (NP_006861) produced in HEK293T cell. | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
11034 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction